Learn More
Medchemexpress LLC Carbonic anhydrase inhibitor 2 | 2758231-43-3 | 98.4% | C12H16N4O6S | 50 MG

Supplier: Medchemexpress LLC HY14284950MG
Carbonic anhydrase inhibitor 2 is a carbonic anhydrase II inhibitor. This compound has been shown to reduce intraocular pressure in glaucomatous rabbits.
- Functions as a carbonic anhydrase II inhibitor
- Reduces intraocular pressure
- High purity of 98.40%
- Provided as a white to off-white solid
- Recommended storage at -20°C for powder, and -80°C in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.