Learn More
Medchemexpress LLC Carbonic anhydrase (isoenzyme) | 9001-03-0 | 110000.00 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1775A10MG
Encompass_Preferred
Carbonic anhydrase isoenzyme is a ubiquitous zinc-containing metalloenzyme found in both prokaryotes and eukaryotes. It functions to catalyze the reversible conversion of carbon dioxide into bicarbonate and protons. This enzyme is utilized in research related to various conditions such as cancer, glaucoma, obesity, and epilepsy.
  • Ubiquitous zinc-containing metalloenzyme
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons
  • Suitable for research in cancer, glaucoma, obesity, and epilepsy
  • Enzyme activity: ≥3000 U/mg solid
Catalog No. 50-004-24089