Learn More
Medchemexpress LLC Carbonic Anhydrase V | 10UG
SDP
Supplier:  Medchemexpress LLC HYP7676510UG
Encompass_Preferred
Despite its name the carbonic anhydrase VIII (CA8) protein lacks carbonic anhydrase catalytic activity This distinguishes CA8 from its typical functions related to carbonic anhydrase an enzyme known for its ability to catalyze the reversible hydration of carbon dioxide Carbonic Anhydrase VIII/CA8 Protein Mouse (His) is the recombinant mouse-derived Carbonic Anhydrase VIII/CA8 protein expressed by E coli with C-His labeled tag
Catalog No. 50-002-93743