Learn More
Cayman Chemical CarboplatIn-d4 1mg
SDP
Supplier:  Cayman Chemical 345061 MG
Encompass_Preferred
An internal standard for the quantification of carboplatin by GC- or LC-MS
Catalog No. 50-295-0262