Learn More
Medchemexpress LLC Carboplatin-d4 | 41575-94-4 | 99.0% | 375.28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17393S5MG
Encompass_Preferred
Carboplatin-d4 is the deuterium labeled Carboplatin. It is a DNA synthesis inhibitor that binds to DNA, inhibits replication and transcription, and induces cell death. This compound is a derivative of CDDP and a potent anti-cancer agent, intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84581