Learn More
Medchemexpress LLC Carbopol 941 polymer | 9007-20-9 | MFCD01675509 | 95.0% | Approximately 1,250,000 g/mol | (C3H4O2)n | 100 G
SDP
Supplier:  Medchemexpress LLC HYW250721C100G
Encompass_Preferred
A crosslinked polyacrylic acid polymer used as a rheology modifier and suspending agent for gels, lotions, and hydro-alcoholic formulations. Supplied as a white powder; typical use levels are 0.1-0.6 wt% for thickening and stabilization.
  • Crosslinked polyacrylic acid powder for rheology modification and thickening.
  • Produces high-viscosity gels and stabilizes emulsions and suspensions.
  • Suitable for clear gels, lotions, and hydro-alcoholic systems.
  • Low recommended use level of 0.1-0.6 wt% (as supplied).
  • White, free-flowing powder for easy dispersion.
  • Provides stable emulsion and suspension performance at low viscosities.
Catalog No. 50-002-80646