Learn More
TARGETMOL CHEMICALS INC CARBOPROST 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T222855MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboprost (15(S)-15-Methyl Prostaglandin F2(alpha)) is a synthetic prostaglandin analogue of PGF2(alpha) which has the effect of oxytocic. Carboprost is commonly used to restore uterine tone. purity: 99%
Catalog No. 50-258-0967