Learn More
Medchemexpress LLC Carboprost tromethamine | 58551-69-2 | 99.85% | C25H47NO8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA019550MG
Encompass_Preferred
Carboprost tromethamine is a synthetic 15-methyl analogue of prostaglandin F2α. It effectively promotes uterine contraction and significantly reduces bleeding during and after delivery. It is for research use only.
  • Solid appearance
  • White to off-white color
  • Shipped at room temperature in continental US; may vary elsewhere
  • Sealed storage at 4°C, away from moisture (solid)
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
  • Soluble in DMSO and H2O (100 mg/mL)
Catalog No. 50-004-33569