Learn More
Medchemexpress LLC Carboprost tromethamine | 58551-69-2 | 99.9% | 489.64 | C25H47NO8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYA019510MG
Encompass_Preferred
Carboprost tromethamine is a synthetic 15-methyl analogue of prostaglandin F2α that acts as a potent uterotonic agent, promoting uterine contraction and reducing postpartum hemorrhage. Supplied for research use, the compound is provided in a 10 mg quantity and should be handled according to safety data guidelines.
  • High purity (99.9%).
  • Molecular formula C25H47NO8; molecular weight 489.64 g/mol.
  • Acts as a prostaglandin receptor agonist with uterotonic activity.
  • Suitable for in vitro and preclinical research applications.
  • Available in multiple pack sizes to support varied experimental needs.
Catalog No. 50-002-98787