Learn More
Apexbio Technology LLC Carboxine 5234-68-4 200mg
SDP
Supplier:  Apexbio Technology LLC C6944200
Encompass_Preferred
Carboxine (5234-68-4) is a small-molecule inhibitor targeting mitochondrial succinate dehydrogenase (SDH) It is designed to inhibit SDH activity thereby interfering with fungal cellular respiration pathways Carboxine exerts its biological activity primarily through inhibition of mitochondrial SDH In in vitro studies Carboxine demonstrates inhibitory efficacy with IC50 values typically in a micromolar range depending on fungal species and assay conditions Based on these pharmacological properties Carboxine holds research potential in studying mitochondrial metabolism energy production pathways fungicidal resistance mechanisms and assessment of biochemical impacts of SDH inhibition within fungal cells
Catalog No. 50-000-90153