Learn More
Medchemexpress LLC Carboxy-PEG4-phosphonic acid | 1623791-69-4 | 96.0% | C11H23O9P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141302100MG
Encompass_Preferred
Carboxy-PEG4-phosphonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds with two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other the protein of interest. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
Catalog No. 50-003-54023