Learn More
Apexbio Technology LLC Carboxy-PTIO, potassium salt 50mg
Supplier:  Apexbio Technology LLC B644550
Encompass_Preferred
Carboxy-PTIO potassium salt (CAS 148819-94-7) is a small molecule recognized as a nitric oxide (NO) scavenger It reacts rapidly with NO converting it to nitrogen oxides and thereby reducing bioavailable NO levels In lipopolysaccharide-induced rat models administration of Carboxy-PTIO potassium salt has been shown to attenuate hypotension and decrease the incidence of endotoxic shock via NO clearance This compound is widely employed in studies investigating NO-related signaling pathways and the pathophysiological roles of aberrant NO production
Catalog No. 50-000-92224