Learn More
Medchemexpress LLC Carboxyatractyloside (dipotassium) | 33286-30-5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN252210MM/1ML
Encompass_Preferred
Carboxyatractyloside dipotassium is a diterpenoid isolated from *Xanthium* plants. It functions as an ADP/ATP carrier inhibitor, specifically targeting mitochondrial ADP/ATP transport. This compound is known to promote reactive oxygen species (ROS) production, induce Ca2+ release, and lead to mitochondrial dysfunction.
  • Inhibits ADP/ATP carrier.
  • Specifically targets mitochondrial ADP/ATP transport.
  • Promotes reactive oxygen species (ROS) production.
  • Induces Ca2+ release.
  • Contributes to mitochondrial dysfunction.
  • Utilized in studies on lethargy, weakness, and epileptic seizures in rats.
Catalog No. 50-004-21070