Learn More
Medchemexpress LLC Carboxybetaine metha 100mg
SDP
Supplier:  Medchemexpress LLC HYW110584100MG
Encompass_Preferred
Carboxybetaine methacrylate is an amphoteric ionic compound with excellent properties such as good hydrophilicity antistatic property and biocompatibility Carboxybetaine methacrylate can be used to prepare tissue engineering scaffolds drug carriers and so on[1][2]
Catalog No. 50-002-05466