Learn More
Abcam 5(6)-Carbo x yeosin, 100MG
Supplier:  Abcam AB274879100MG
Encompass_Preferred
MW 1383.8 Da, Purity >98%. Brominated analog of carboxyfluorescein that is a photosensitizer and singlet oxygen generator (19 times greater than fluorescein). Specific PMCA (plasma membrane calcium pump) inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-244