Learn More
LUNA NANOTECH INC Carboxyl Coated Magnetic Beads, 500Nm, 10 ML
Supplier: LUNA NANOTECH INC MGBCOOH1010
500 nm diameter magnetic beads with Carboxyl (COOH) functionalized surface.
Features:
- Conjugation ready: COOH groups on the magnetic beads can be reacted with NH2 groups to covalently load proteins, small molecules, or DNA / RNA onto the beads.
- High monodispersity.
- Stable in high salt conditions.
- Fast response to a magnet.
Applications:
- Magnetic separation of DNA, RNA, proteins, antibodies, cells and small molecules from complex samples.
- NH2 containing proteins, small molecules, DNA / RNA can be covalently attached to the carboxyl groups on the bead surface through EDC/NHS reactions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.