Learn More
TARGETMOL CHEMICALS INC CARBOXYLESTERASE-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T775245MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboxylesterase-IN-2 is a potent Carboxylesterase Notum inhibitor (IC50 <= 10 nM).Carboxylesterase-IN-2 has potential for the study of cancer diseases. purity: 99%
Catalog No. 50-257-9138