Learn More
Medchemexpress LLC Carboxylesterase-IN-3 | 2764748-92-5 | 98.27% | C11H6Cl2N4OS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY142689100MG
Encompass_Preferred
Carboxylesterase-IN-3 (compound 4y) is a potent inhibitor of Carboxylesterase Notum with an IC50 less than or equal to 10 nM. Notum is a negative regulator of Wnt signaling, which acts through the hydrolysis of a palmitoleoylate ester, essential for Wnt activity. Carboxylesterase-IN-3 has the potential for the research of cancer disease.
  • Potent inhibitor of Carboxylesterase Notum
  • IC50 less than or equal to 10 nM
  • Negative regulator of Wnt signaling
  • Potential for research of cancer disease
Catalog No. 50-003-56443