Learn More
Medchemexpress LLC Carboxylesterase-IN-3 | 2764748-92-5 | 98.3% | C11H6Cl2N4OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14268950MG
Encompass_Preferred
Carboxylesterase-IN-3 is a potent inhibitor of Carboxylesterase Notum, demonstrating an IC50 of 10 nM or less. Notum is recognized as a negative regulator of Wnt signaling, a process crucial for Wnt activity, which it mediates through the hydrolysis of a palmitoleoylate ester. This compound shows potential for research applications in cancer studies.
  • Potent inhibitor of carboxylesterase notum.
  • Inhibits Wnt signaling by targeting notum.
  • Aids in cancer disease research.
  • High purity of 98.3%.
  • Available in solid and solution forms for research flexibility.
Catalog No. 50-003-56201