Learn More
Sigma Aldrich Fine Chemicals Biosciences Carboxymethyl-beta-cyclode1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 219061G
Encompass
Carboxymethyl--cyclodextrin a negatively charged derivative of -cyclodextrin is generally used in the chiral analysis of neutral and basic analytes on account of its ability to exhibit counter-current mobility.
Catalog No. 50-290-0831