Learn More
Medchemexpress LLC Carboxymethyl-β-cyclodextrin | 218269-34-2 | 99.7% | 1541.24 | 10 G
SDP
Supplier:  Medchemexpress LLC HY12639310G
Encompass_Preferred
Carboxymethyl-β-cyclodextrin is a cationic cyclodextrin utilized as a carrier for various drugs. It demonstrates high affinity and selectivity in binding nonsteroidal anti-inflammatory drugs.
  • Used as a drug carrier.
  • Binds nonsteroidal anti-inflammatory drugs with high affinity.
  • Exhibits selectivity for its binding targets.
Catalog No. 50-004-38270