Learn More
Medchemexpress LLC Carboxymethyl-β-cyclodextrin | 218269-34-2 | 99.7% | 1541.24 | 5 G
Supplier: Medchemexpress LLC HY1263935G
Carboxymethyl-β-cyclodextrin is a cationic cyclodextrin utilized as a drug carrier. It has demonstrated high affinity and selectivity for binding nonsteroidal anti-inflammatory drugs (NSAIDs).
- Used as a drug carrier
- Binds nonsteroidal anti-inflammatory drugs (NSAIDs) with high affinity and selectivity