Learn More
TOKU-E Carbenicillin Disodium, CulturePure, 4800-94-6, 1 g
Supplier:  TOKU-E C1071G
Encompass_Preferred
carboxypenicillin antibiotic, selection antibiotic, C17H16N2Na2O6S. Formulated for cell culture, plant tissue culture.
Catalog No. 50-255-1811