Learn More
Medchemexpress LLC Carboxypeptidase A1/A2/B Antibody (YA2486) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8274150UL
Encompass_Preferred
Carboxypeptidase A1/A2/B Antibody (YA2486) is a monoclonal antibody designed for specific detection of Carboxypeptidase A1/A2/B proteins. This Rabbit-derived, non-conjugated IgG antibody is affinity purified for high specificity.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets Carboxypeptidase A1/A2/B
  • Affinity purified
  • Observed band size: 44 kDa
  • Predicted band size: 47 kDa
Catalog No. 50-004-43615