Learn More
Worthington Biochemical Corporation CARBOXYPEPTIDASE B 10MG
SDP
Supplier:  Worthington Biochemical Corporation LS005301
Encompass
Carboxypeptidase B, 10mg, MW: 34500 daltons, pH: 7.9, Chromatographically Purified, -20degreeC Storage Temp, Source: Porcine Pancreas
Catalog No. NC9098116