Learn More
Medchemexpress LLC Carboxypeptidase B2/CPB2 protein, rat (HEK293, His) | >95.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7544910UG
Encompass_Preferred
Recombinant rat carboxypeptidase B2 (CPB2) expressed in HEK293 cells with a C-terminal His tag. This glycosylated protease cleaves C-terminal arginine and lysine residues and is provided as a purified protein for biochemical characterization, enzymatic assays, antibody generation, and structural studies.
  • Expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity >95% by reducing SDS-PAGE.
  • Glycosylated; apparent molecular weight ~55-75 kDa.
  • Suitable for biochemical assays, enzymatic activity studies, antibody production, and structural assays.
  • Available in microgram and milligram quantities.
Catalog No. 50-002-91316