Learn More
Foxx Life Sciences EZBio® 20L (5 GAL) Gamma Sterilized Double Bagged Autoclavable Polypropylene (PP) with 83mm (83B) VersaCap® Closure
Supplier:  Foxx Life Sciences 5155H51FLS
Encompass_Preferred
Foxx Life Sciences EZBio® Gamma Sterile Polypropylene Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.Polypropylene carboys are made from a USP Class VI, FDA Grade, autoclavable material.EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 26.5L Max Capacity.
Catalog No. 50-172-0679