Learn More
Medchemexpress LLC Carcainium chloride | 1042-42-8 | MFCD00864735 | 99.5% | 347.84 g/mol | C18H22ClN3O2 | 1 ML

Supplier: Medchemexpress LLC HY106372A10MM 1ML
Carcainium chloride is a quaternary derivative of lidocaine used as a research compound in pharmacology and electrophysiology. It is supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and has reported antitussive activity.
- Quaternary derivative of lidocaine.
- Reported antitussive activity.
- High purity (~99.5%).
- Available as solid or 10 mM solution in DMSO (1 mL).
- Molecular formula C18H22ClN3O2; molecular weight 347.84 g/mol.
- Intended for research use in pharmacology and electrophysiology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.