Learn More
Thomas Scientific Cardboard Freezer Box, Purple
SDP
Supplier:  Thomas Scientific 1188Y27
Encompass_Preferred
Unique color band design Available in 5 bright color options (blue, purple, pink, green & orange) For use with 81 & 100 place dividers Ample white space and marking area allows for labeling sample identification 5-1/4 x 5-1/4 x 2 inches; fits most standard freezer racks
Catalog No. 50-145-7468