Learn More
Research Products International Corp Cardboard Storage Box w/ Lid & Divider, 100 Capacity
SDP
Supplier:  Research Products International Corp 181037
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator use. Boxes contain a cardboard cell partition to help hold popular size tubes. Partitions accept tubes with a diameter less than 11.65 mm.
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-197-8701