Learn More
Apexbio Technology LLC Cardiogreen,100mg CAS# 3599-32-4
SDP
Supplier:  Apexbio Technology LLC B8315100
Encompass_Preferred
Cardiogreen, also known as indocyanine green, is a nontoxic tricarbocyanine dye, with a peak spectral absorption at 790 nm. Cardiogreen administered intravenously rapidly binds to its principle carrier, plasma protein, and then becomes confined to the vascular space. Cardiogreen has been commonly used in medical diagnostics for measuring cardiac output, liver blood flow, and hepatic function, as well as for ophthalmic angiography. In addition, cardiogreen can also be used as a photosensitizer in the photodynamic therapy (PDT) for treating diseases like periodontal infections. References:1. "Indocyanine green solution". NIH. National Cancer Institute. Retrieved 20 August 2021.2. Gharesi S, Pourhajibagher M, Chiniforush N, et al. Effect of photodynamic therapy based on indocyanine green on expression of apoptosis-related genes in human gingival fibroblast cells. Photodiagnosis and Photodynamic Therapy , 2017, 19: 33-36. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0130