Learn More
Medchemexpress LLC Cardiolipin (heart, bovine) sodium | 383907-10-6 | MFCD11974098 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1342385MG
Encompass_Preferred
Cardiolipin (heart, bovine) sodium is a research-grade mitochondrial phospholipid extracted from bovine heart tissue. Supplied as a white to off-white solid sodium salt with high purity, it is used as a reagent in biochemical assays, mitochondrial and lipid research, and for coating microplates in immunoassays. Store sealed, away from moisture and light; follow solvent-specific storage recommendations.
Catalog No. 50-002-22949