Learn More
Medchemexpress LLC Carfilzomib-d8 | 1537187-53-3 | 98.7% | 727.96 g/mol | C40H49D8N5O7 | 500 UG

Supplier: Medchemexpress LLC HY10455S500UG
Carfilzomib-d8 is a deuterium-labeled analog of carfilzomib, an irreversible proteasome inhibitor used in research. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is suitable for studies of proteasome inhibition in cell models.
- Deuterium-labeled internal standard for quantitative mass spectrometry and NMR.
- Irreversible proteasome inhibitor activity; reported IC50 ≈ 5 nM in ANBL-6 and RPMI 8226 cells.
- Molecular formula C40H49D8N5O7; molecular weight 727.96 g/mol.
- Typical purity around 98.7% (batch dependent).
- Storage: protect from light; in solvent store at -80 °C up to 6 months, -20 °C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.