Learn More
Medchemexpress LLC Carfilzomib-d8 | 1537187-53-3 | >99.0% | 728.0 g/mol | C40H49D8N5O7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10455S1MG
Encompass_Preferred
Carfilzomib-d8 is the octa-deuterated form of carfilzomib supplied for research and analytical use as an internal standard. It enables accurate quantification of carfilzomib in biological and pharmaceutical samples by GC- or LC-MS, with high isotopic enrichment and analytical-grade purity.
  • Octa-deuterated internal standard suitable for LC- or GC-MS quantitation.
  • High isotopic enrichment and purity for minimal analytical interference.
  • Suitable for quantification in biological matrices and pharmacokinetic studies.
  • Provided in small, pre-weighed quantities compatible with analytical workflows.
  • Molecular formula C40H49D8N5O7 and formula weight 728.0 g/mol for reference.
Catalog No. 50-002-44433