Learn More
Selleck Chemical LLC Carfilzomib (PR-171) 5mg 868540-17-4
SDP
Supplier:  Selleck Chemical LLC S28535MG
Encompass_Preferred
Carfilzomib (PR-171) is an irreversible proteasome inhibitor with IC50 of <5 nM in ANBL-6 cells, displayed preferential in vitro inhibitory potency against the ChT-L activity in the ?5 subunit, but little or no effect on the PGPH and T-L activities. Carfilzomib activates prosurvival autophagy and induces cell apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7948