Learn More
Medchemexpress LLC Carglumic Acid | 1188-38-1 | MFCD00047874 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB071110MM/1ML
Encompass_Preferred
Carglumic acid (N-Carbamyl-L-glutamic acid) is a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator. It is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.
  • Functional analogue of N-acetylglutamate (NAG).
  • Activates carbamoyl phosphate synthetase 1 (CPS1).
  • Used to treat acute and chronic hyperammonemia.
  • Suppresses cell viability in pancreatic, breast, hepatoma, and lung cancer cell lines.
  • Significantly inhibits tumor growth in orthotopic cancer models.
Catalog No. 50-004-07934