Learn More
Medchemexpress LLC Carglumic acid (standard) | 1188-38-1 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0711R
Encompass_Preferred
Carglumic Acid (Standard) is the analytical standard of Carglumic Acid, intended for research and analytical applications. Carglumic acid, a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.
  • Used as a reference standard for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS techniques
Catalog No. 50-004-50799