Learn More
Medchemexpress LLC Cariprazine-d6 | 1308278-67-2 | 98.7% | 433.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14763S5MG
Encompass_Preferred
Cariprazine-d6 is a deuterium-labeled version of Cariprazine, an antipsychotic agent. It demonstrates high affinity for dopamine D3 (Ki of 0.085 nM) and D2 (Ki of 0.49 nM) receptors, along with a moderate affinity for the 5-HT1A receptor (Ki of 2.6 nM).
  • Deuterium labeled
  • High affinity for D3 and D2 receptors
  • Moderate affinity for 5-HT1A receptor
  • Suitable as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-59740