Learn More
Medchemexpress LLC Carmofur | 61422-45-5 | 99.95% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0182100MG
Encompass_Preferred
Carmofur (HCFU) is a rat recombinant acid ceramidase inhibitor with an IC50 of 29 nM. It also acts as a protease inhibitor for SARS-CoV-2 main protease (Mpro), fatty acid amide hydrolase (FAAH), and N-acylethanolamine acid amidase (NAAA). This compound exhibits anti-cancer, anti-inflammatory, and anti-virus activities, making it suitable for studying COVID-19 and acute lung injury (ALI).
  • Inhibits acid ceramidase (IC50: 29 nM)
  • Acts as a protease inhibitor for SARS-CoV-2 Mpro
  • Inhibits fatty acid amide hydrolase (FAAH)
  • Inhibits N-acylethanolamine acid amidase (NAAA)
  • Exhibits anti-cancer activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-virus activity
  • Used for studying COVID-19 and acute lung injury (ALI)
Catalog No. 50-004-07389