Learn More
Apexbio Technology LLC Carmofur 61422-45-5 100mg
SDP
Supplier:  Apexbio Technology LLC A2548100
Encompass_Preferred
Carmofur is a fluorouracil derivative that functions as an anticancer agent by inhibiting thymidylate synthetase thereby interfering with DNA synthesis and cell proliferation As a cytostatic compound it is used primarily in oncology research for in vitro studies investigating tumor growth inhibition pathways Research demonstrates Carmofur s cytotoxic activity in various cancer cell lines with reported IC50 values typically ranging from approximately 5 to 50 M depending on the cell type and culture conditions It is commonly applied in cellular assays including viability assays apoptosis pathway analysis and mechanistic studies of fluorouracil resistance providing insight into targeted pharmacological interventions and chemotherapeutic response mechanisms
Catalog No. 50-000-63555