Learn More
Medchemexpress LLC Carmoterol hydrochloride | 137888-11-0 | 98.7% | 404.89 g/mol | C21H25ClN2O4 | 10MM 1ML

Supplier: Medchemexpress LLC HY1246310MM 1ML
Carmoterol hydrochloride is a potent, selective, long-acting β2-adrenoceptor agonist supplied for research use. It is available as solid samples and as a ready-to-use 10 mM solution in DMSO (1 mL), and is intended for pharmacology and respiratory research including bronchodilation, asthma, and COPD models.
- High purity suitable for research use.
- Available as solid and as a 10 mM solution in DMSO.
- Molecular weight 404.89 g/mol.
- CAS number 137888-11-0 for unambiguous identification.
- Supplied as the hydrochloride salt for improved solubility.
- Suitable for β2-adrenergic receptor pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.