Learn More
Medchemexpress LLC Carmoterol (hydrochloride) | 137888-11-0 | 98.7% | 404.89 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12463
Encompass_Preferred
Carmoterol hydrochloride is a highly potent, selective, and long-acting β2-adrenoceptor agonist with a pEC50 of 10.19. It exhibits 53 times higher affinity for β2-adrenoceptors compared to β1-adrenoceptors. This compound is suitable for research related to asthma and chronic obstructive pulmonary disease (COPD).
  • Highly potent, selective, and long-acting β2-adrenoceptor agonist.
  • 53 times higher affinity for β2-adrenoceptors than for β1-adrenoceptors.
  • Can be used for the research of asthma and chronic obstructive pulmonary disease (COPD).
  • High purity: 98.73%.
Catalog No. 50-004-39246