Learn More
TARGETMOL CHEMICALS INC CARMUSTINE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3091100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carmustine (bis-chloroethylnitrosourea) is a cell-cycle phase nonspecific alkylating antineoplastic agent. purity: 99%
Catalog No. 50-257-7005