Learn More
Medchemexpress LLC HY-13585 50mg , Carmustine CAS:154-93-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1358550MG
Encompass_Preferred
Medchemexpress, HY-13585 50mg Carmustine CAS:154-93-8 Formula:C5H9Cl2N3O2 DNA Alkylator Purity:98% Carmustine is an antitumor chemotherapeutic agent, which works by akylating DNA and RNA . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3724