Learn More
Cayman Chemical CarnosIc AcId 10mg
SDP
Supplier:  Cayman Chemical 8982010 MG
Encompass_Preferred
An antioxidant derived from rosemary and common sage that inhibits lipid peroxidation and scavenges peroxyl radicals; cytotoxic towards various human cancer cell lines, reducing cell viability to 13-30% at 6.25 UG/ml
Catalog No. 50-289-6330