Learn More
Selleck Chemical LLC Carnosic acid S3838-5mg
SDP
Supplier:  Selleck Chemical LLC S38385MG
Encompass_Preferred
Carnosic acid (Salvin) is a phenolic diterpene endowed with antioxidative and antimicrobial properties It is increasingly used within food nutritional health and cosmetics industries
Catalog No. 50-311-0378