Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | 332.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0644100MG
Encompass_Preferred
Carnosic acid is an orally active compound and phenolic diterpene found in Lamiaceae plants, including *Rosmarinus officinalis L.* and *Salvia officinalis*. It inhibits lipid absorption, oxidative stress, and inflammation. It also suppresses cell proliferation and exhibits antibacterial activity.
  • Inhibits oxidative stress
  • Reduces inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • May have therapeutic potential in different types of cancer
  • Demonstrates neuroprotective effects in experimental models of neurodegenerative diseases
Catalog No. 50-003-89069