Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | 332.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0644
Encompass_Preferred
Carnosic acid is an orally active lipid absorption inhibitor. It has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity. This phenolic diterpene is enriched in the leaves of Lamiaceae plants, such as Rosmarinus officinalis L. and Salvia officinalis, and has shown therapeutic potential in various cancers and neuroprotective effects through NRF2/ARE pathway activation.
  • Orally active lipid absorption inhibitor
  • Inhibits oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Neuroprotective effects
Catalog No. 50-003-90332