Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | 332.43 | 50 MG

Supplier: Medchemexpress LLC HYN0644R50MG
Carnosic acid (Standard) is an analytical standard used for research and analytical applications. It exhibits inhibition of oxidative stress and inflammation, suppresses cell proliferation, and demonstrates antibacterial activity. This reference standard is widely utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
- Purity: 98.43%
- Appearance: Solid
- Color: Light yellow to yellow
- Structure classification: Terpenoids, Diterpenoids, Phenols, Polyphenols
- Initial source: Plants Leguminosae Paramacrolobium coeruleum (Taub.) J.Léonard, Plants Labiatae
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.