Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | 332.43 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN0644500MG
Encompass_Preferred
Carnosic acid is an orally active lipid absorption inhibitor. It has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity. It shows therapeutic potential in various cancers and neuroprotective effects in neurodegenerative disease models.
  • Inhibits oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Potential therapeutic agent in various cancers
  • Neuroprotective effects in neurodegenerative disease models
Catalog No. 50-003-89070